Methyl 3-(2-Pyridyl)propanoate

Lieferant: Apollo Scientific

OR470609-5G
APOSOR470609-5GEA 300 EUR
APOSOR470609-5G
Methyl 3-(2-Pyridyl)propanoate
Methyl 3-(2-Pyridyl)propanoate
Formel: C₉H₁₁NO₂
Molecular Weight: 165,189 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18803099
CAS-Nummer: 28819-26-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR