3-(3-Pyridyl)propanamide

Lieferant: Apollo Scientific

OR470611-5G
APOSOR470611-5GEA 997 EUR
APOSOR470611-5G
3-(3-Pyridyl)propanamide
3-(3-Pyridyl)propanamide
Formel: C₈H₁₀N₂O
Molecular Weight: 150,178 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00956246
CAS-Nummer: 84199-99-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR