Ethyl 3-(2-Pyridyl)propiolate

Lieferant: Apollo Scientific

OR470613-5G
APOSOR470613-5GEA 1220 EUR
APOSOR470613-5G
Ethyl 3-(2-Pyridyl)propiolate
Ethyl 3-(2-Pyridyl)propiolate
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381104
CAS-Nummer: 66869-70-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR