Ethyl 3-(3-Pyridyl)propiolate

Lieferant: Apollo Scientific

OR470614-5G
APOSOR470614-5GEA 1220 EUR
APOSOR470614-5G
Ethyl 3-(3-Pyridyl)propiolate
Ethyl 3-(3-Pyridyl)propiolate
Formel: C₁₀H₉NO₂
Molecular Weight: 175,184 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01073808
CAS-Nummer: 59608-03-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR