Methyl 3-(4-Pyridyl)propiolate

Lieferant: Apollo Scientific

OR470615-5G
APOSOR470615-5GEA 1220 EUR
APOSOR470615-5G
Methyl 3-(4-Pyridyl)propiolate
Methyl 3-(4-Pyridyl)propiolate
Formel: C₉H₇NO₂
Molecular Weight: 161,157 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18205099
CAS-Nummer: 78584-33-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR