3-(3-Pyridyl)propiolic acid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-(Pyridin-3-yl)propiolic acid
OR470617-5G
APOSOR470617-5GEA
1420
EUR
APOSOR470617-5G
3-(3-Pyridyl)propiolic acid
3-(3-Pyridyl)propiolic acid
Formel:
C₈H₅NO₂ Molecular Weight: 147,131 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02177466 CAS-Nummer: 59608-01-4 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...