4-(2-Pyridyl)-3-butyn-1-ol

Lieferant: Apollo Scientific

Synonyme: 4-(Pyridin-2-yl)but-3-yn-1-ol

OR470620-5G
APOSOR470620-5GEA 517 EUR
APOSOR470620-5G
4-(2-Pyridyl)-3-butyn-1-ol
4-(2-Pyridyl)-3-butyn-1-ol
Formel: C₉H₉NO
Molecular Weight: 147,174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD18384524
CAS-Nummer: 395652-44-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR