1-Acetyl-3-piperidineacetic acid

Lieferant: Apollo Scientific

OR470622-1G
APOSOR470622-1GEA 499 EUR
APOSOR470622-1G
1-Acetyl-3-piperidineacetic acid
1-Acetyl-3-piperidineacetic acid
Formel: C₉H₁₅NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD14582888
CAS-Nummer: 169253-07-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR