3-(Boc-amino)-4-bromo-6-chloropyridazine

Lieferant: Apollo Scientific

OR470623-1G
APOSOR470623-1GEA 397 EUR
APOSOR470623-1G
3-(Boc-amino)-4-bromo-6-chloropyridazine
3-(Boc-amino)-4-bromo-6-chloropyridazine
Formel: C₉H₁₁BrClN₃O₂
Molecular Weight: 308,56 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23381126
CAS-Nummer: 1427475-32-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR