3-Amino-5-bromo-4-methoxypyridine

Lieferant: Apollo Scientific

OR470624-1G
APOSOR470624-1GEA 243 EUR
APOSOR470624-1G
3-Amino-5-bromo-4-methoxypyridine
3-Amino-5-bromo-4-methoxypyridine
Formel: C₆H₇BrN₂O
Molecular Weight: 203.037 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11100691
CAS-Nummer: 31872-78-3

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR