2-Chloro-N-(4-pyridyl)acetamide
Lieferant: Apollo Scientific
OR470625-5G
APOSOR470625-5GEA
804
EUR
APOSOR470625-5G
2-Chloro-N-(4-pyridyl)acetamide
2-Chloro-N-(4-pyridyl)acetamide
Formel:
C₇H₇ClN₂O Molecular Weight: 170,596 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00996250 CAS-Nummer: 80650-46-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...