5-(Diethylamino)-3,6-dimethylpyrazine-2-carboxylic acid

Lieferant: Apollo Scientific

OR470633-1G
APOSOR470633-1GEA 804 EUR
APOSOR470633-1G
5-(Diethylamino)-3,6-dimethylpyrazine-2-carboxylic acid
5-(Diethylamino)-3,6-dimethylpyrazine-2-carboxylic acid
Formel: C₁₁H₁₇N₃O₂
Molecular Weight: 223,272 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24335246
CAS-Nummer: 1434128-60-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR