1-(3,5-Dichlorophenyl)-3-(ethoxycarbonyl)urea

Lieferant: Apollo Scientific

OR470634-1G
APOSOR470634-1GEA 804 EUR
APOSOR470634-1G
1-(3,5-Dichlorophenyl)-3-(ethoxycarbonyl)urea
1-(3,5-Dichlorophenyl)-3-(ethoxycarbonyl)urea
Formel: C₁₀H₁₀Cl₂N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00135183
CAS-Nummer: 199741-78-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR