2-(3-Nitrophenyl)-1,3-dioxolane

Lieferant: Apollo Scientific

OR470637-25G
APOSOR470637-25GEA 377 EUR
APOSOR470637-25G
2-(3-Nitrophenyl)-1,3-dioxolane
2-(3-Nitrophenyl)-1,3-dioxolane
Formel: C₉H₉NO₄
Molecular Weight: 195,172 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020899
CAS-Nummer: 6952-67-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR