3'-Amino-4'-methylacetophenone

Lieferant: Apollo Scientific

Synonyme: 1-(3-Amino-4-methylphenyl)ethanone

OR470638-1G
APOSOR470638-1GEA 134 EUR
APOSOR470638-1G
3'-Amino-4'-methylacetophenone
3'-Amino-4'-methylacetophenone
Formel: C₉H₁₁NO
Molecular Weight: 149,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07787585
CAS-Nummer: 17071-24-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR