4-Bromo-3-(dibromomethyl)benzonitrile

Lieferant: Apollo Scientific

OR470639-5G
APOSOR470639-5GEA 1120 EUR
APOSOR470639-5G
4-Bromo-3-(dibromomethyl)benzonitrile
4-Bromo-3-(dibromomethyl)benzonitrile
Formel: C₈H₄Br₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD24335261
CAS-Nummer: 160313-49-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR