Methyl 4-bromo-2-(4-methyl-1-piperazinyl)benzoate

Lieferant: Apollo Scientific

OR470640-5G
APOSOR470640-5GEA 713 EUR
APOSOR470640-5G
Methyl 4-bromo-2-(4-methyl-1-piperazinyl)benzoate
Methyl 4-bromo-2-(4-methyl-1-piperazinyl)benzoate
Formel: C₁₃H₁₇BrN₂O₂
Molecular Weight: 313,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24335260
CAS-Nummer: 1434128-45-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR