Methyl 2-(4-Bromo-2-nitrophenyl)-2-(dimethoxyphosphoryl)acetate

Lieferant: Apollo Scientific

OR470642-5G
APOSOR470642-5GEA 1330 EUR
APOSOR470642-5G
Methyl 2-(4-Bromo-2-nitrophenyl)-2-(dimethoxyphosphoryl)acetate
Methyl 2-(4-Bromo-2-nitrophenyl)-2-(dimethoxyphosphoryl)acetate
Formel: C₁₁H₁₃BrNO₇P
Molecular Weight: 382,101 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24335257
CAS-Nummer: 1094851-34-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR