Ethyl 5-[(Diphenylmethylene)amino]-2-pyrimidinecarboxylate
Lieferant: Apollo Scientific
OR470643-1G
APOSOR470643-1GEA
713
EUR
APOSOR470643-1G
Ethyl 5-[(Diphenylmethylene)amino]-2-pyrimidinecarboxylate
Ethyl 5-[(Diphenylmethylene)amino]-2-pyrimidinecarboxylate
Formel:
C₂₀H₁₇N₃O₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD24335252 CAS-Nummer: 1434128-44-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...