Methyl 5-bromo-3-(2-chloroacetyl)-2-hydroxybenzoate

Lieferant: Apollo Scientific

OR470646-5G
APOSOR470646-5GEA 1330 EUR
APOSOR470646-5G
Methyl 5-bromo-3-(2-chloroacetyl)-2-hydroxybenzoate
Methyl 5-bromo-3-(2-chloroacetyl)-2-hydroxybenzoate
Formel: C₁₀H₈BrClO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD24335248
CAS-Nummer: 1434128-48-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR