2-Amino-4-chloro-7-methoxyquinazoline
Lieferant: Apollo Scientific
OR470647-1G
APOSOR470647-1GEA
713
EUR
APOSOR470647-1G
2-Amino-4-chloro-7-methoxyquinazoline
2-Amino-4-chloro-7-methoxyquinazoline
Formel:
C₉H₈ClN₃O Molecular Weight: 209,632 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18378816 CAS-Nummer: 181871-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...