2-Amino-4-chloro-6-methoxyquinazoline

Lieferant: Apollo Scientific

OR470648-1G
APOSOR470648-1GEA 713 EUR
APOSOR470648-1G
2-Amino-4-chloro-6-methoxyquinazoline
2-Amino-4-chloro-6-methoxyquinazoline
Formel: C₉H₈ClN₃O
Lagertemperatur: Raumtemperatur
MDL: MFCD18378817
CAS-Nummer: 1379296-48-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR