4-(1-Piperidylmethyl)phenylboronic acid
Lieferant: Apollo Scientific
Synonyme:
4-(Piperidin-1-ylmethyl)phenylboronic acid
OR470649-1G
APOSOR470649-1GEA
601
EUR
APOSOR470649-1G
4-(1-Piperidylmethyl)phenylboronic acid
4-(1-Piperidylmethyl)phenylboronic acid
Formel:
C₁₂H₁₈BNO₂ Molecular Weight: 219.088 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06801706 CAS-Nummer: 1200434-84-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...