4-(1-Pyrrolidylmethyl)phenylboronic acid

Lieferant: Apollo Scientific
Achtung

OR470650-1G
APOSOR470650-1GEA 601 EUR
APOSOR470650-1G
4-(1-Pyrrolidylmethyl)phenylboronic acid
4-(1-Pyrrolidylmethyl)phenylboronic acid
Formel: C₁₁H₁₆BNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06801708
CAS-Nummer: 1036991-20-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR