N-Methoxy-N-methylpyrazine-2-carboxamide

Lieferant: Apollo Scientific

OR470651-5G
APOSOR470651-5GEA 1330 EUR
APOSOR470651-5G
N-Methoxy-N-methylpyrazine-2-carboxamide
N-Methoxy-N-methylpyrazine-2-carboxamide
Formel: C₇H₉N₃O₂
Molecular Weight: 167,165 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06804574
CAS-Nummer: 317335-26-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR