3-(Hydroxymethyl)-1-methyl-2-pyrrolidinone
Lieferant: Apollo Scientific
Synonyme:
3-(Hydroxymethyl)-1-methylpyrrolidin-2-one
OR470654-1G
APOSOR470654-1GEA
128
EUR
APOSOR470654-1G
3-(Hydroxymethyl)-1-methyl-2-pyrrolidinone
3-(Hydroxymethyl)-1-methyl-2-pyrrolidinone
Formel:
C₆H₁₁NO₂ Molecular Weight: 129,157 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD18816870 CAS-Nummer: 577780-05-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...