3-Ethoxypyridin-2(1H)-one
Lieferant: Apollo Scientific
OR470655-1G
APOSOR470655-1GEA
804
EUR
APOSOR470655-1G
3-Ethoxypyridin-2(1H)-one
3-Ethoxypyridin-2(1H)-one
Formel:
C₇H₉NO₂ Molecular Weight: 139,152 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22554050 CAS-Nummer: 909854-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...