Ethyl 4,4-Dimethyl-2-oxopyrrolidine-3-carboxylate

Lieferant: Apollo Scientific

OR470656-1G
APOSOR470656-1GEA 764 EUR
APOSOR470656-1G
Ethyl 4,4-Dimethyl-2-oxopyrrolidine-3-carboxylate
Ethyl 4,4-Dimethyl-2-oxopyrrolidine-3-carboxylate
Formel: C₉H₁₅NO₃
Molecular Weight: 185,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD12828110
CAS-Nummer: 90609-07-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR