5-Amino-2-(4-nitrophenyl)pyrimidine

Lieferant: Apollo Scientific

OR470657-1G
APOSOR470657-1GEA 896 EUR
APOSOR470657-1G
5-Amino-2-(4-nitrophenyl)pyrimidine
5-Amino-2-(4-nitrophenyl)pyrimidine
Formel: C₁₀H₈N₄O₂
Molecular Weight: 216,196 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03145364
CAS-Nummer: 131548-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR