4-(5-Amino-2-pyrimidyl)phenol

Lieferant: Apollo Scientific

OR470658-1G
APOSOR470658-1GEA 896 EUR
APOSOR470658-1G
4-(5-Amino-2-pyrimidyl)phenol
4-(5-Amino-2-pyrimidyl)phenol
Formel: C₁₀H₉N₃O
Molecular Weight: 187,198 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11180875
CAS-Nummer: 1094311-62-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR