5-Amino-2-(2-pyridyl)pyrimidine

Lieferant: Apollo Scientific

OR470659-1G
APOSOR470659-1GEA 896 EUR
APOSOR470659-1G
5-Amino-2-(2-pyridyl)pyrimidine
5-Amino-2-(2-pyridyl)pyrimidine
Formel: C₉H₈N₄
Molecular Weight: 172.187 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11180855
CAS-Nummer: 1094246-40-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR