5-Amino-2-(4-bromophenyl)pyrimidine
Lieferant: Apollo Scientific
OR470660-1G
APOSOR470660-1GEA
896
EUR
APOSOR470660-1G
5-Amino-2-(4-bromophenyl)pyrimidine
5-Amino-2-(4-bromophenyl)pyrimidine
Formel:
C₁₀H₈BrN₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11180884 CAS-Nummer: 84610-01-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...