Methyl 3-(4-Methyl-1-piperazinyl)propanoate

Lieferant: Apollo Scientific

OR470661-5G
APOSOR470661-5GEA 804 EUR
APOSOR470661-5G
Methyl 3-(4-Methyl-1-piperazinyl)propanoate
Methyl 3-(4-Methyl-1-piperazinyl)propanoate
Formel: C₉H₁₈N₂O₂
Molecular Weight: 186,251 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08062551
CAS-Nummer: 33544-40-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR