4-(Boc-amino)-1-(3-pyridyl)piperidine

Lieferant: Apollo Scientific

OR470662-5G
APOSOR470662-5GEA 1180 EUR
APOSOR470662-5G
4-(Boc-amino)-1-(3-pyridyl)piperidine
4-(Boc-amino)-1-(3-pyridyl)piperidine
Formel: C₁₅H₂₃N₃O₂
Molecular Weight: 277,362 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24470614
CAS-Nummer: 1013920-63-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR