2-Chloro-2'-hydroxy-3'-methylacetophenone
Lieferant: Apollo Scientific
OR470663-1G
APOSOR470663-1GEA
601
EUR
APOSOR470663-1G
2-Chloro-2'-hydroxy-3'-methylacetophenone
2-Chloro-2'-hydroxy-3'-methylacetophenone
Formel:
C₉H₉ClO₂ Molecular Weight: 184,62 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD13172653 CAS-Nummer: 75717-51-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...