1-Methyl-5-nitro-1,2,3-triazole

Lieferant: Apollo Scientific

OR470666-1G
APOSOR470666-1GEA 601 EUR
APOSOR470666-1G
1-Methyl-5-nitro-1,2,3-triazole
1-Methyl-5-nitro-1,2,3-triazole
Formel: C₃H₄N₄O₂
Molecular Weight: 128,089 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26045236
CAS-Nummer: 1204736-25-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR