4-Amino-2-chloro-5-[(trimethylsilyl)ethynyl]benzonitrile

Lieferant: Apollo Scientific

OR470667-1G
APOSOR470667-1GEA 804 EUR
APOSOR470667-1G
4-Amino-2-chloro-5-[(trimethylsilyl)ethynyl]benzonitrile
4-Amino-2-chloro-5-[(trimethylsilyl)ethynyl]benzonitrile
Formel: C₁₂H₁₃ClN₂Si
Molecular Weight: 248,784 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26045235
CAS-Nummer: 1425933-51-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR