5-(Dibenzylamino)-2-methyl-2-pentanol

Lieferant: Apollo Scientific

OR470668-1G
APOSOR470668-1GEA 499 EUR
APOSOR470668-1G
5-(Dibenzylamino)-2-methyl-2-pentanol
5-(Dibenzylamino)-2-methyl-2-pentanol
Formel: C₂₀H₂₇NO
Molecular Weight: 297,434 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26045234
CAS-Nummer: 1455037-19-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR