Ethyl 2-(4,6-Dichloro-5-pyrimidyl)acetate

Lieferant: Apollo Scientific

OR470670-1G
APOSOR470670-1GEA 804 EUR
APOSOR470670-1G
Ethyl 2-(4,6-Dichloro-5-pyrimidyl)acetate
Ethyl 2-(4,6-Dichloro-5-pyrimidyl)acetate
Formel: C₈H₈Cl₂N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD13195351
CAS-Nummer: 1095822-21-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR