2-(Boc-amino)-5-bromo-4-(hydroxymethyl)pyridine

Lieferant: Apollo Scientific

OR470671-1G
APOSOR470671-1GEA 519 EUR
APOSOR470671-1G
2-(Boc-amino)-5-bromo-4-(hydroxymethyl)pyridine
2-(Boc-amino)-5-bromo-4-(hydroxymethyl)pyridine
Formel: C₁₁H₁₅BrN₂O₃
Molecular Weight: 303.152 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26045232
CAS-Nummer: 1823773-02-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR