Methyl 1-(tert-Butyl)-5-(2,4-dichlorophenyl)pyrazole-3-carboxylate

Lieferant: Apollo Scientific

OR470673-1G
APOSOR470673-1GEA 397 EUR
APOSOR470673-1G
Methyl 1-(tert-Butyl)-5-(2,4-dichlorophenyl)pyrazole-3-carboxylate
Methyl 1-(tert-Butyl)-5-(2,4-dichlorophenyl)pyrazole-3-carboxylate
Formel: C₁₅H₁₆Cl₂N₂O₂
Molecular Weight: 327,206 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26045230
CAS-Nummer: 2006277-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR