Ethyl 2-[4-(Methylsulphonamido)phenyl]acetate

Lieferant: Apollo Scientific

OR470675-5G
APOSOR470675-5GEA 713 EUR
APOSOR470675-5G
Ethyl 2-[4-(Methylsulphonamido)phenyl]acetate
Ethyl 2-[4-(Methylsulphonamido)phenyl]acetate
Formel: C₁₁H₁₅NO₄S
Molecular Weight: 257,306 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26045229
CAS-Nummer: 58827-89-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR