1H-Imidazo[4,5-b]pyrazine

Lieferant: Apollo Scientific

OR470676-1G
APOSOR470676-1GEA 178 EUR
APOSOR470676-1G
1H-Imidazo[4,5-b]pyrazine
1H-Imidazo[4,5-b]pyrazine
Formel: C₅H₄N₄
Molecular Weight: 120,112 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08669465
CAS-Nummer: 273-94-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR