1-Cbz-3-[Boc(cyclopropyl)amino]piperidine

Lieferant: Apollo Scientific

OR470678-5G
APOSOR470678-5GEA 1210 EUR
APOSOR470678-5G
1-Cbz-3-[Boc(cyclopropyl)amino]piperidine
1-Cbz-3-[Boc(cyclopropyl)amino]piperidine
Formel: C₂₁H₃₀N₂O₄
Molecular Weight: 374,474 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26404523
CAS-Nummer: 1823267-91-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR