Methyl-3-amino-3-(2-thienyl)propanoate

Lieferant: Apollo Scientific

OR470681-1G
APOSOR470681-1GEA 306 EUR
APOSOR470681-1G
Methyl-3-amino-3-(2-thienyl)propanoate
Methyl-3-amino-3-(2-thienyl)propanoate
Formel: C₈H₁₁NO₂S
Molecular Weight: 185.25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03001286
CAS-Nummer: 188812-40-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR