(S)-2-(3-Chloro-2-hydroxypropoxy)isoindoline-1,3-dione

Lieferant: Apollo Scientific

OR470688-5G
APOSOR470688-5GEA 916 EUR
APOSOR470688-5G
(S)-2-(3-Chloro-2-hydroxypropoxy)isoindoline-1,3-dione
(S)-2-(3-Chloro-2-hydroxypropoxy)isoindoline-1,3-dione
Formel: C₁₁H₁₀ClNO₄
Molecular Weight: 255,654 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26523287
CAS-Nummer: 272447-40-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR