6-(Boc-aminomethyl)-1,2,3-benzotriazole

Lieferant: Apollo Scientific

OR470691-5G
APOSOR470691-5GEA 1330 EUR
APOSOR470691-5G
6-(Boc-aminomethyl)-1,2,3-benzotriazole
6-(Boc-aminomethyl)-1,2,3-benzotriazole
Formel: C₁₂H₁₆N₄O₂
Molecular Weight: 248,281 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26523285
CAS-Nummer: 1823790-58-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR