Methyl 2-Acetamido-3-(4-pyridyl)propanoate
Lieferant: Apollo Scientific
OR470692-1G
APOSOR470692-1GEA
1120
EUR
APOSOR470692-1G
Methyl 2-Acetamido-3-(4-pyridyl)propanoate
Methyl 2-Acetamido-3-(4-pyridyl)propanoate
Formel:
C₁₁H₁₄N₂O₃ Molecular Weight: 222,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD26523296 CAS-Nummer: 138808-56-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...