Methyl 2-Acetamido-3-(3-thienyl)propanoate

Lieferant: Apollo Scientific

OR470694-1G
APOSOR470694-1GEA 1120 EUR
APOSOR470694-1G
Methyl 2-Acetamido-3-(3-thienyl)propanoate
Methyl 2-Acetamido-3-(3-thienyl)propanoate
Formel: C₁₀H₁₃NO₃S
Molecular Weight: 227,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD26523295
CAS-Nummer: 1227732-30-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR