2-Acetamido-3-(1-imidazolyl)propanoic acid

Lieferant: Apollo Scientific

OR470695-1G
APOSOR470695-1GEA 916 EUR
APOSOR470695-1G
2-Acetamido-3-(1-imidazolyl)propanoic acid
2-Acetamido-3-(1-imidazolyl)propanoic acid
Formel: C₈H₁₁N₃O₃
Molecular Weight: 197,191 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20364913
CAS-Nummer: 848555-92-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR